Merck
  • Overexpression of succinyl-CoA synthase for poly (3-hydroxybutyrate-co-3-hydroxyvalerate) production in engineered Escherichia coli BL21(DE3).
Overexpression of succinyl-CoA synthase for poly (3-hydroxybutyrate-co-3-hydroxyvalerate) production in engineered Escherichia coli BL21(DE3).
Journal of applied microbiology (2015-06-26)
S K Bhatia, D-H Yi, H-J Kim, J-M Jeon, Y-H Kim, G Sathiyanarayanan, H M Seo, J H Lee, J-H Kim, K Park, C J Brigham, Y-H Yang
ABSTRAKT
This study aims to increase the 3-hydroxyvalerate (3HV) fraction in poly(3-hydroxybutyrate-co-3-hydroxyvalerate) [P(HB-co-HV)] using succinyl-CoA synthase. Escherichia coli YH090, a polyhydroxyalkonate (PHA)-producing strain, was further engineered for overexpression of succinyl-CoA synthase genes (sucCD), and examined for P(HB-co-HV) copolymer production in the presence of various precursor molecules using mixture analysis. Glycerol, succinate and propionate were screened as important factors for controlling intracellular PHA accumulation and monomer composition. Glycerol concentrations exerted the greatest influence on the overall biomass concentration and the intracellular PHA content, while propionate concentrations in the presence of succinate influenced the 3HV content of the copolymer. Mixture analysis also demonstrated that the engineered strain has the capacity to accumulate up to 80% of its cell dry weight (CDW) as PHA with a variable fraction of 3HV monomer (maximum of 72 wt %) depending on the controlled conditions. Propionate is the principal precursor for 3HV monomer in P(HB-co-HV) biopolymer and its utilization requires conversion to propionyl-CoA. Engineered E. coli YHY99, overexpressing sucCD genes, leads to an increase of the succinyl-CoA pool, which enhances the conversion rate of propionate by providing a CoA supply to other acyltransferase enzymes that have a role in propionate utilization. Engineered E. coli YHY99 was able to utilize propionate with a 4·5-fold increase in rate, as compared to the control strain, and resulted in the synthesis of a copolymer with high 3HV monomer content.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium propionate, ≥99.0%
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium propionate, FCC
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside