Merck
Changes in the hemolytic activity of Candida species by common electrolytes.
BMC microbiology (2015-08-25)
Lei Wan, Gang Luo, Haibin Lu, Dongying Xuan, Hong Cao, Jincai Zhang
ABSTRAKT
Hemolysins are crucial virulence factors which help pathogens to survive and persist in the host. This study investigated whether common electrolytes will affect the hemolysins of Candida species. The hemolysins from 25 Candida isolates were investigated using a plate specially designed for Candida species in the presence of three electrolytes, CaCl2, NaCl and KCl, at different concentrations. The hemolytic activity was determined after 48 h and the hemolytic index was calculated. All three electrolytes caused a decrease in the hemolytic activity. Significant differences existed between varying concentrations of NaCl, while no significant differences existed for the CaCl2 and KCl groups. Additionally, the peripheral hemolytic index was highly correlated with the hemolytic index (r = 0.656, p < 0.001). Our findings indicate that electrolytes reduce hemolysis by Candida species and a correlation exists between the peripheral hemolytic index and the translucent hemolytic index.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O