Merck
Interleukin-1β protects astrocytes against oxidant-induced injury via an NF-κB-dependent upregulation of glutathione synthesis.
Glia (2015-04-17)
Yan He, Nicole A Jackman, Trista L Thorn, Valarie E Vought, Sandra J Hewett
ABSTRAKT
Astrocytes produce and export the antioxidant glutathione (GSH). Previously, we found that interleukin-1β (IL-1β) enhanced the expression of astrocyte system xc (-) , the transporter that delivers the rate-limiting substrate for GSH synthesis-cyst(e)ine. Herein, we demonstrate directly that IL-1β mediates a time-dependent increase in extracellular GSH levels in cortical astrocyte cultures, suggesting both enhanced synthesis and export. This increased GSH production was blocked by inhibition of nuclear factor-κB (NF-κB) activity but not by inhibition of p38 MAPK. To determine whether this increase could provide protection against oxidative stress, the oxidants tert-butyl hydroperoxide (tBOOH) and ferrous sulfate (FeSO4 ) were employed. IL-1β treatment prevented the increase in reactive oxygen species produced in astrocytes following tBOOH exposure. Additionally, the toxicity induced by tBOOH or FeSO4 exposure was significantly attenuated following treatment with IL-1β, an effect reversed by concomitant exposure to l-buthionine-S,R-sulfoximine (BSO), which prevented the IL-1β-mediated rise in GSH production. IL-1β failed to increase GSH or to provide protection against t-BOOH toxicity in astrocyte cultures derived from IL-1R1 null mutant mice. Overall, our data indicate that under certain conditions IL-1β may be an important stimulus for increasing astrocyte GSH production, and potentially, total antioxidant capacity in brain, via an NF-κB-dependent process.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
Sodium Fluoride Solution
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, ≥98%, lyophilized powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
SAFC
L-Glutathione oxidized