Merck
Hepatic metallothioneins in molecular responses to cobalt, zinc, and their nanoscale polymeric composites in frog Rana ridibunda.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2015-05-20)
Halina Falfushynska, Lesya Gnatyshyna, Olga Fedoruk, Natalia Mitina, Alexander Zaichenko, Oksana Stoliar, Rostyslav Stoika
ABSTRAKT
Despite numerous studies suggesting a dramatic decline of amphibians, the biochemical mechanisms of adaptation in these animals to polluted environment are poorly studied. The aim of this study was to elucidate the ability to release cobalt (Co) and zinc (Zn) from their nanoscale complexes (NCs) derived from the polymeric substance of N-vinylpyrrolidone (PS) in the liver of amphibian (Rana ridibunda). Frog males were subjected to 14days exposure to waterborne Co(2+) (50μg/L), Zn(2+) (100μg/L), as well as corresponding concentrations of Co-NC, Zn-NC or PS. Main attention was paid to MT's interrelations with indices of stress and toxicity. Only Co(2+) and Zn(2+) caused elevation of the correspondent metal in MTs. Co(2+) caused down-regulation of cathepsin D activity, while Zn(2+), Zn-NC and the PS up-regulated this activity. Zn(2+) provoked 1.6 times increase of metal-bounded form of the MT (MT-Me), while all other exposures caused the elevation of the ratio of MT total protein concentration (MT-SH) and concentrations of the MT-Me and/or immunoreactive (MTi) form (up to ~10 times) accompanied by a decrease in the levels of oxyradicals. The increased DNA fragmentation and down-regulation of caspase-3 activity in relation to the redox state of glutathione and/or lactate/pyruvate were shown at all exposures. These data indicate the vulnerability of the redox state of cellular thiols and inability to release Co and Zn from NCs in frog's liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
2-Vinylpyridine, 97%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)