Merck
Characterization of secretomes provides evidence for adipose-derived mesenchymal stromal cells subtypes.
Stem cell research & therapy (2015-11-13)
Natalia Kalinina, Daria Kharlampieva, Marina Loguinova, Ivan Butenko, Olga Pobeguts, Anastasia Efimenko, Luidmila Ageeva, George Sharonov, Dmitry Ischenko, Dmitry Alekseev, Olga Grigorieva, Veronika Sysoeva, Ksenia Rubina, Vassiliy Lazarev, Vadim Govorun
ABSTRAKT
This study was aimed at deciphering the secretome of adipose-derived mesenchymal stromal cells (ADSCs) cultured in standard and hypoxic conditions to reveal proteins, which may be responsible for regenerative action of these cells. Human ADSCs were isolated from 10 healthy donors and cultured for 3-4 passages. Cells were serum deprived and cell purity was assessed using multiple cell surface markers. Conditioned media was collected and analyzed using LC-MS with a focus on characterizing secreted proteins. Purity of the ADSC assessed as CD90+/CD73+/CD105+/CD45-/CD31- cells was greater than 99 % and viability was greater than 97 %. More than 600 secreted proteins were detected in conditioned media of ADSCs. Of these 100 proteins were common to all cultures and included key molecules involved in tissue regeneration such as collagens and collagen maturation enzymes, matrix metalloproteases, matricellular proteins, macrophage-colony stimulating factor and pigment epithelium derived factor. Common set of proteins also included molecules, which contribute to regenerative processes but were not previously associated with ADSCs. These included olfactomedin-like 3, follistatin-like 1 and prosaposin. In addition, ADSCs from the different subjects secreted proteins, which were variable between different cultures. These included proteins with neurotrophic activities, which were not previously associated with ADSCs, such as mesencephalic astrocyte-derived neurotrophic factor, meteorin and neuron derived neurotrophic factor. Hypoxia resulted in secretion of 6 proteins, the most prominent included EGF-like repeats and discoidin I-like domains 3, adrenomedullin and ribonuclease 4 of RNase A family. It also caused the disappearance of 8 proteins, including regulator of osteogenic differentiation cartilage-associated protein. Human ADSCs with CD90+/CD73+/CD105+/CD45-/CD31-/PDGFRβ+/NG2+/CD146+(-) immunophenotype secrete a large array of proteins, the most represented group is comprised of extracellular matrix components. Number of secreted proteins is largely unaffected by prolonged hypoxia. Variability in the secretion of several proteins from cultured ADSCs of individual subjects suggests that these cells exist as a heterogeneous population containing functionally distinct subtypes, which differ in numbers between donors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
SAFC
Iodoacetamide