Skip to Content
MilliporeSigma

Skip To

AABH97CD7DB1

AA BLOCKS, INC.

Mecoprop 2-ethylhexyl ester

Synonym(s):

2-ethylhexyl 2-(4-chloro-2-methylphenoxy)propanoate

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Ships in 21 days - from Aldrich Partner.
$114.40
100 mg
Ships in 21 days - from Aldrich Partner.
$750.10

About This Item

Linear Formula:
C18H27O3Cl1
CAS Number:
Molecular Weight:
326.86
MDL number:
Sign in to see your pricing

Vendor SKU: AA00FASC-10MG

$114.40


Ships in 21 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H27ClO3/c1-5-7-8-15(6-2)12-21-18(20)14(4)22-17-10-9-16(19)11-13(17)3/h9-11,14-15H,5-8,12H2,1-4H3

Inchi Key

NWTKQOOQEPXMMW-UHFFFAOYSA-N

Country of Origin

US


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AABH9BD93229ALNH9A9FA1F8TA9H98771BA5
description

-

description

-

description

-

description

-

Inchi Key

NWTKQOOQEPXMMW-UHFFFAOYSA-N

Inchi Key

FZXUXQSWWHWPNU-UHFFFAOYSA-N

Inchi Key

-

Inchi Key

-

Country of Origin

US

Country of Origin

US

Country of Origin

US

Country of Origin

CN

Inchi Code

1S/C18H27ClO3/c1-5-7-8-15(6-2)12-21-18(20)14(4)22-17-10-9-16(19)11-13(17)3/h9-11,14-15H,5-8,12H2,1-4H3

Inchi Code

1S/C18H27ClO3/c1-5-6-7-8-9-14(3)21-18(20)15(4)22-17-11-10-16(19)12-13(17)2/h10-12,14-15H,5-9H2,1-4H3

Inchi Code

1S/C10H11ClO3/c1-6-5-8(11)3-4-9(6)14-7(2)10(12)13/h3-5,7H,1-2H3,(H,12,13)

Inchi Code

1S/C10H11ClO3/c1-6-5-8(11)3-4-9(6)14-7(2)10(12)13/h3-5,7H,1-2H3,(H,12,13)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library