Skip to Content
MilliporeSigma

BDPH9BCE13A2

Bide Pharmatech

2-Chloro-1H-indole-3-carbaldehyde

Synonym(s):

2-chloro-1H-indole-3-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1N1Cl1
CAS Number:
Molecular Weight:
179.61
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6ClNO/c10-9-7(5-12)6-3-1-2-4-8(6)11-9/h1-5,11H

Inchi Key

XYSSNBNFOBVMAU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 2-Chloro-1H-indole-3-carbaldehyde


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library