Skip to Content
MilliporeSigma

Skip To

COM448635526

Combi-Blocks, Inc.

5-Fluoro-2-(2-methoxyethoxy)pyridine

Synonym(s):

5-fluoro-2-(2-methoxyethoxy)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mg
Ships in 7 days - from Aldrich Partner.
$65.00
1 g
Ships in 7 days - from Aldrich Partner.
$169.00
5 g
Ships in 7 days - from Aldrich Partner.
$676.00

About This Item

Linear Formula:
C8H10O2N1F1
CAS Number:
Molecular Weight:
171.17
MDL number:

Vendor SKU: PY-1876-250MG

$65.00


Ships in 7 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10FNO2/c1-11-4-5-12-8-3-2-7(9)6-10-8/h2-3,6H,4-5H2,1H3

Inchi Key

DFSYOWPLGXRNLV-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library