Skip to Content
MilliporeSigma

Skip To

APREST91908

PrEST Antigen FBXO41

Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

Fbx41, KIAA1940

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160702


recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 27 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

ARREVFESTSFQGKEQAAGPSPAAPHLLHHHHHHAPLAHFPGDLVPASLPCEELAEPGLVPAAAARYALREIEIPLGELFARKSVAS

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FBXO41(150726)

General description

Recombinant protein fragment of Human FBXO41 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.    

Application

Blocking agent and positive assay control using corresponding antibodies.

Physical form

Solution in phosphate-buffered saline and 1M Urea, pH 7.4

Preparation Note

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.

Other Notes

Corresponding Antibody HPA058386.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters