Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $617.00 |
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
$617.00
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
51 kDa
species reactivity
human
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MSL3L2(151507)
Immunogen
Synthetic peptide directed towards the middle region of human MSL3L2
Application
Anti-MSL3L2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.
Biochem/physiol Actions
Male-specific lethal 3 homolog (Drosophila) pseudogene 1 (MSL3P1; MSL3L2) forms a multisubunit human histone acetyltransferase complex (hMSL) that is involved in H4 acetylation at lysine 16. Acetylationof H4 is represses the expression of some genes and cell cycle arrest in G2/M phase.
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Other Notes
Synthetic peptide located within the following region: EKLSPSLRLLNPSRPQSTESQSTSGEPATPKRRKAEPQAVQSLRRSSPHT
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.