Skip to Content
MilliporeSigma

Skip To

PLA0262

Rabbit anti-Dcp2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DCP2 decapping enzyme homolog, DCP2 decapping enzyme homolog (S. cerevisiae), M(7)GpppN-mRNA hydrolase, NUDT20, Nucleoside diphosphate-linked moiety X motif 20, Nudix motif 20, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
$576.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-

$576.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-15 μg/mg, western blot: 1:1,000-1:5,000

accession no.

NP_689837.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Dcp2(167227)

Immunogen

The epitope recognized by PLA0262 maps to a region between residue 370 and 420 of human mRNA-decapping enzyme 2 using the numbering given in entry NP_689837.2 (GeneID 167227).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

mRNA-decapping enzyme 2 (Dcp2) is required for both normal mRNA turnover and nonsense-mediated mRNA decay. Dcp2 plays a role in replication-dependent histone mRNA degradation. It removes the 7-methyl guanine cap structure from mRNA molecules, yielding a 5′-phosphorylated mRNA fragment and 7m-GDP [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/Q8IU60].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D6319HPA057676HPA041252
description

Powered by Bethyl Laboratories, Inc.

description

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

human

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

immunoprecipitation (IP): 5-15 μg/mg, western blot: 1:1,000-1:5,000

technique(s)

indirect immunofluorescence: suitable, western blot: 2-4 μg/mL using lysaes of K-562 and Rat1 cells, indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde-fixed NIH-3T3 celle

technique(s)

-

technique(s)

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters