Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
96 wells | Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery) | $660.00 |
About This Item
sample type serum
sample type plasma
$660.00
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 82 pg/mL
standard curve range: 0.082-20 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... H60a(15101)
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description for serum, plasma and cell culture supernatant |
| species reactivity mouse | species reactivity mouse | species reactivity mouse | species reactivity mouse |
| Gene Information mouse ... H60a(15101) | Gene Information - | Gene Information mouse ... Ifnz(319146) | Gene Information mouse ... Il4(16189) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <10% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| input sample type cell culture supernatant(s) | input - | input sample type plasma | input sample type plasma |
| detection method colorimetric | detection method - | detection method colorimetric | detection method colorimetric |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
10 - Combustible liquids
Hazard Classifications
Resp. Sens. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




