Skip to Content
MilliporeSigma
vibrant-m

MCDB 153 Medium

vibrant-m
Synonym(s):
Molecular, Cellular, and Development Biology 153 medium

Products

Product Category
Brand
Physical Form
Quality Segment
Technique