MilliporeSigma
  • Loss of RASGRP1 in humans impairs T-cell expansion leading to Epstein-Barr virus susceptibility.
Loss of RASGRP1 in humans impairs T-cell expansion leading to Epstein-Barr virus susceptibility.
EMBO molecular medicine (2017-12-29)
Sarah Winter, Emmanuel Martin, David Boutboul, Christelle Lenoir, Sabah Boudjemaa, Arnaud Petit, Capucine Picard, Alain Fischer, Guy Leverger, Sylvain Latour
ABSTRACT
Inherited CTPS1, CD27, and CD70 deficiencies in humans have revealed key factors of T-lymphocyte expansion, a critical prerequisite for an efficient immunity to Epstein-Barr virus (EBV) infection. RASGRP1 is a T-lymphocyte-specific nucleotide exchange factor known to activate the pathway of MAP kinases (MAPK). A deleterious homozygous mutation in RASGRP1 leading to the loss RASGRP1 expression was identified in two siblings who both developed a persistent EBV infection leading to Hodgkin lymphoma. RASGRP1-deficient T cells exhibited defective MAPK activation and impaired proliferation that was restored by expression of wild-type RASGRP1. Similar defects were observed in T cells from healthy individuals when RASGRP1 was downregulated. RASGRP1-deficient T cells also exhibited decreased CD27-dependent proliferation toward CD70-expressing EBV-transformed B cells, a crucial pathway required for expansion of antigen-specific T cells during anti-EBV immunity. Furthermore, RASGRP1-deficient T cells failed to upregulate CTPS1, an important enzyme involved in DNA synthesis. These results show that RASGRP1 deficiency leads to susceptibility to EBV infection and demonstrate the key role of RASGRP1 at the crossroad of pathways required for the expansion of activated T lymphocytes.
Sigma-Aldrich
Anti-RasGRP1 Antibody, clone 10.1, clone 10.1, from mouse
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution