MilliporeSigma
  • Rapid method for identification of macrophages in suspension by acid alpha-naphthyl acetate esterase activity.
Rapid method for identification of macrophages in suspension by acid alpha-naphthyl acetate esterase activity.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1983-07-01)
D L Ennist, K H Jones
ABSTRACT
A supravital staining procedure for the identification of macrophages in cell suspension using a modification of a standard cytochemical assay for alpha-naphthyl acetate esterase (ANAE) activity is described. Macrophages are stained an intense red-brown after 5 min incubation in a buffer using ANAE as the substrate and hexazonium pararosaniline as the coupler for the azo dye. There is close agreement in the number of ANAE-positive cells found and the number of macrophages identified in smears by morphological criteria, by phagocytosis, and by the presence of Fc receptors. Therefore, this stain provides a quick, inexpensive method to estimate the number of macrophages present in suspensions of lymphocytic tissues from rats and mice.
Sigma-Aldrich
1-Naphthyl acetate, ≥98% (C)
Sigma-Aldrich
3,4-Dichloroisocoumarin, serine protease inhibitor