MilliporeSigma
  • Defining the antibody cross-reactome directed against the influenza virus surface glycoproteins.
Defining the antibody cross-reactome directed against the influenza virus surface glycoproteins.
Nature immunology (2017-02-14)
Raffael Nachbagauer, Angela Choi, Ariana Hirsh, Irina Margine, Sayaka Iida, Aldo Barrera, Marcela Ferres, Randy A Albrecht, Adolfo García-Sastre, Nicole M Bouvier, Kimihito Ito, Rafael A Medina, Peter Palese, Florian Krammer
ABSTRACT
Infection with influenza virus induces antibodies to the viral surface glycoproteins hemagglutinin and neuraminidase, and these responses can be broadly protective. To assess the breadth and magnitude of antibody responses, we sequentially infected mice, guinea pigs and ferrets with divergent H1N1 or H3N2 subtypes of influenza virus. We measured antibody responses by ELISA of an extensive panel of recombinant glycoproteins representing the viral diversity in nature. Guinea pigs developed high titers of broadly cross-reactive antibodies; mice and ferrets exhibited narrower humoral responses. Then, we compared antibody responses after infection of humans with influenza virus H1N1 or H3N2 and found markedly broad responses and cogent evidence for 'original antigenic sin'. This work will inform the design of universal vaccines against influenza virus and can guide pandemic-preparedness efforts directed against emerging influenza viruses.
Sigma-Aldrich
Anti-Human IgG (Fab specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Guinea Pig IgG Antibody, HRP conjugate, Chemicon®, from goat