MilliporeSigma
Micropropagation of tulip: production of virus-free stock plants.
Methods in molecular biology (Clifton, N.J.) (2010-01-26)
Małgorzata Podwyszyńska, Dariusz Sochacki
ABSTRACT
We describe here a new tulip micropropagation method based on the cyclic shoot multiplication in presence of the thidiazuron (TDZ), which enables the production of virus-free stock plants, speeds up breeding, and provides new genotypes for the market. In our novel protocol, cyclic shoot multiplication can be performed for 2-3 years by using TDZ instead of other cytokinins, as 6-benzylaminopurine (BAP) and N(6)-(-isopentyl)adenine (2iP). It makes possible to produce 500-2,000 microbulbs from one healthy plant. There are six main stages of tulip micropropagation. Stage 0 is the selection of true-to-type and virus-free plants, confirmed by ELISA. Fragments of flower stems isolated from bulbs are used as initial explants. Shoot multiplication is based on the regeneration of adventitious shoots, which are sub-cultured every 8 weeks. In the Stage 3, the specially prepared shoots are induced by low temperature treatment to form bulbs which finally develop on a sucrose-rich medium at 20 degrees C. Bulbs are then dried for 6 weeks and rooted in vivo. The number of multiplication subcultures should be limited to 5-10 cycles in order to lower the risk of mutation. Virus indexing should be repeated 3-4 times, at the initial stage and then during shoot multiplication. Genetic stability of micropropagated shoots can be confirmed using molecular markers.
Sigma-Aldrich
Thidiazuron, suitable for plant cell culture, BioReagent
Supelco
Thidiazuron, PESTANAL®, analytical standard