MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Tyrosinase inhibitory components from the seeds of Cassia tora.
Archives of pharmacal research (2018-05-04)
Ga Young Lee, Byoung Ok Cho, Jae Young Shin, Seon Il Jang, In Sook Cho, Hyo Young Kim, Ji Su Park, Chong Woon Cho, Jong Seong Kang, Jang Hoon Kim, Young Ho Kim
ABSTRACT
Ten compounds (1-10) isolated from the seeds of Cassia tora were evaluated for tyrosinase inhibition. Compounds 3, 4, and 7 inhibited tyrosinase enzymatic activity in a dose-dependent manner, with IC50 values of 3.0 ± 0.8, 7.0 ± 0.4, and 9.2 ± 3.4 μM, respectively. Kinetic analyses revealed a mechanism consistent with competitive inhibition. In silico molecular docking showed that compounds 3 and 4 docked in the active site of tyrosinase, whereas 7 interacted with Ala246 and Val248 at outside of the active site, and His244 and Glu256 at inside. Additionally, compounds 3, 4, and 7 suppressed melanogenesis in α-MSH-treated B16F10 melanoma cells at a concentration of 10 μM.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid