MilliporeSigma
  • A new diphosphine-carbonyl complex of ruthenium: an efficient precursor for C-C and C-N bond coupling catalysis.
A new diphosphine-carbonyl complex of ruthenium: an efficient precursor for C-C and C-N bond coupling catalysis.
Dalton transactions (Cambridge, England : 2003) (2018-07-18)
Aparajita Mukherjee, David A Hrovat, Michael G Richmond, Samaresh Bhattacharya
ABSTRACT
Reaction of 1,2-bis(diphenylphosphino)benzene (dppbz) with [{Ru(CO)2Cl2}n] affords [Ru(dppbz)(CO)2Cl2], where the two carbonyls are mutually cis and the two chlorides are trans. The molecular structure of [Ru(dppbz)(CO)2Cl2], has been determined by X-ray crystallography, and the stability of the different available stereoisomers has been computationally evaluated. [Ru(dppbz)(CO)2Cl2] has been found to serve as an excellent pre-catalyst for catalytic Suzuki-type C-C coupling and Buchwald-type C-N coupling reactions.
Sigma-Aldrich
1,2-Bis(diphenylphosphino)benzene, 97%
Sigma-Aldrich
1,3-Propylene sulfite, 99%