MilliporeSigma
  • Vaccinia virus late transcription is activated in vitro by cellular heterogeneous nuclear ribonucleoproteins.
Vaccinia virus late transcription is activated in vitro by cellular heterogeneous nuclear ribonucleoproteins.
The Journal of biological chemistry (2001-09-08)
C F Wright, B W Oswald, S Dellis
ABSTRACT
Vaccinia virus gene expression is temporally regulated, and three gene classes have been identified: early, intermediate, and late. Several virus-encoded proteins and an activity designated VLTF-X are required for maximum transcription in vitro of a template containing a late promoter. VLTF-X is present in both cytoplasmic and nuclear extracts prepared from uninfected mammalian cells and co-purifies with a late promoter DNA-binding activity. Here, extensive purification of VLTF-X has revealed that heterogeneous nuclear ribonucleoproteins A2/B1 and RBM3 co-purified with in vitro late transcription stimulation. Overexpression and purification of these proteins from Escherichia coli demonstrated that they both complemented for VLTF-X activity in in vitro transcription reactions. These studies identify two host cell factors potentially contributing to poxvirus replication in vivo.
Sigma-Aldrich
Polyuridylic acid–Agarose, lyophilized powder, matrix polyacrylhydrazido-agarose