MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Arabidopsis RNA processing factor SERRATE regulates the transcription of intronless genes.
Arabidopsis RNA processing factor SERRATE regulates the transcription of intronless genes.
eLife (2018-08-29)
Corinna Speth, Emese Xochitl Szabo, Claudia Martinho, Silvio Collani, Sven Zur Oven-Krockhaus, Sandra Richter, Irina Droste-Borel, Boris Macek, York-Dieter Stierhof, Markus Schmid, Chang Liu, Sascha Laubinger
ABSTRACT
Intron splicing increases proteome complexity, promotes RNA stability, and enhances transcription. However, introns and the concomitant need for splicing extend the time required for gene expression and can cause an undesirable delay in the activation of genes. Here, we show that the plant microRNA processing factor SERRATE (SE) plays an unexpected and pivotal role in the regulation of intronless genes. Arabidopsis SE associated with more than 1000, mainly intronless, genes in a transcription-dependent manner. Chromatin-bound SE liaised with paused and elongating polymerase II complexes and promoted their association with intronless target genes. Our results indicate that stress-responsive genes contain no or few introns, which negatively affects their expression strength, but that some genes circumvent this limitation via a novel SE-dependent transcriptional activation mechanism. Transcriptome analysis of a Drosophila mutant defective in ARS2, the metazoan homologue of SE, suggests that SE/ARS2 function in regulating intronless genes might be conserved across kingdoms.
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution