MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Serine 25 phosphorylation inhibits RIPK1 kinase-dependent cell death in models of infection and inflammation.
Serine 25 phosphorylation inhibits RIPK1 kinase-dependent cell death in models of infection and inflammation.
Nature communications (2019-04-17)
Yves Dondelinger, Tom Delanghe, Dario Priem, Meghan A Wynosky-Dolfi, Daniel Sorobetea, Diego Rojas-Rivera, Piero Giansanti, Ria Roelandt, Julia Gropengiesser, Klaus Ruckdeschel, Savvas N Savvides, Albert J R Heck, Peter Vandenabeele, Igor E Brodsky, Mathieu J M Bertrand
ABSTRACT
RIPK1 regulates cell death and inflammation through kinase-dependent and -independent mechanisms. As a scaffold, RIPK1 inhibits caspase-8-dependent apoptosis and RIPK3/MLKL-dependent necroptosis. As a kinase, RIPK1 paradoxically induces these cell death modalities. The molecular switch between RIPK1 pro-survival and pro-death functions remains poorly understood. We identify phosphorylation of RIPK1 on Ser25 by IKKs as a key mechanism directly inhibiting RIPK1 kinase activity and preventing TNF-mediated RIPK1-dependent cell death. Mimicking Ser25 phosphorylation (S > D mutation) protects cells and mice from the cytotoxic effect of TNF in conditions of IKK inhibition. In line with their roles in IKK activation, TNF-induced Ser25 phosphorylation of RIPK1 is defective in TAK1- or SHARPIN-deficient cells and restoring phosphorylation protects these cells from TNF-induced death. Importantly, mimicking Ser25 phosphorylation compromises the in vivo cell death-dependent immune control of Yersinia infection, a physiological model of TAK1/IKK inhibition, and rescues the cell death-induced multi-organ inflammatory phenotype of the SHARPIN-deficient mice.
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-MLKL (Ser345) Antibody, clone 7C6.1, clone 7C6.1, from mouse
Sigma-Aldrich
Anti-MLKL Antibody, clone 3H1, clone 3H1, from rat