MilliporeSigma
  • The Stat3 inhibitor, S3I-201, downregulates lymphocyte activation markers, chemokine receptors, and inflammatory cytokines in the BTBR T+ Itpr3tf/J mouse model of autism.
The Stat3 inhibitor, S3I-201, downregulates lymphocyte activation markers, chemokine receptors, and inflammatory cytokines in the BTBR T+ Itpr3tf/J mouse model of autism.
Brain research bulletin (2019-07-13)
Sheikh F Ahmad, Mushtaq A Ansari, Ahmed Nadeem, Saleh A Bakheet, Ahmed Z Alanazi, Sary Alsanea, Homood M As Sobeai, Mashal M Almutairi, Hafiz M Mahmood, Sabry M Attia
ABSTRACT
Autism is a complex neurodevelopmental disorder with a high incidence rate. It is characterized by deficits in communication, a lack of social skills, cognitive inflexibility, and stereotypical behaviors. Autism has been gradually increasing in children over the past several years, without the existence of an effective treatment. BTBR T+ Itpr3tf/J (BTBR) mice serve as an accepted model to evaluate autistic-like behaviors as they display core behavioral symptoms displayed in autism. Previous findings showed that S3I-201, a selective Stat3 inhibitor, can be used to treat neuroinflammation disorders. Previously, we showed that S3I-201 treatment has therapeutic effects on autism-like behaviors, and Th1/Th17 and regulatory T cells in BTBR mice. The objective of the present study was to further explore the role of S3I-201 in BTBR mice, and this was performed by investigating the effects of S3I-201 treatment on lymphocyte activation markers (CD4+CD25+ and CD4+CD69+), chemokine receptors (CD4+CCR6+, CD4+CCR7+, CD4+CXCR4+, and CD4+CXCR5+), and proinflammatory cytokines (CD4+IL-6+ and CD4+TNF-α+) in the spleen cells of BTBR and C57BL/6 (C57) mice. The mRNA and protein expression levels of CD69, CCR6, CCR7, CXCR4, CXCR5, IL-1β, IL-6, and TNF-α were examined in the brain tissues, and in BTBR mice, a significant decrease in CD25, CD69, CCR6, CCR7, CXCR4, CXCR5, IL-6, and TNF-α producing CD4+ T cells was observed. The present findings suggest that treatment with S3I-201 may be a therapeutic approach to improve immune abnormalities in a subgroup of autistic subjects.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
S3I-201, ≥97% (HPLC)