MilliporeSigma
  • ADAM17-dependent c-MET-STAT3 signaling mediates resistance to MEK inhibitors in KRAS mutant colorectal cancer.
ADAM17-dependent c-MET-STAT3 signaling mediates resistance to MEK inhibitors in KRAS mutant colorectal cancer.
Cell reports (2014-06-17)
Sandra Van Schaeybroeck, Murugan Kalimutho, Philip D Dunne, Robbie Carson, Wendy Allen, Puthen V Jithesh, Keara L Redmond, Takehiko Sasazuki, Senji Shirasawa, Jaine Blayney, Paolo Michieli, Cathy Fenning, Heinz-Josef Lenz, Mark Lawler, Daniel B Longley, Patrick G Johnston
ABSTRACT
There are currently no approved targeted therapies for advanced KRAS mutant (KRASMT) colorectal cancer (CRC). Using a unique systems biology approach, we identified JAK1/2-dependent activation of STAT3 as the key mediator of resistance to MEK inhibitors in KRASMT CRC in vitro and in vivo. Further analyses identified acute increases in c-MET activity following treatment with MEK inhibitors in KRASMT CRC models, which was demonstrated to promote JAK1/2-STAT3-mediated resistance. Furthermore, activation of c-MET following MEK inhibition was found to be due to inhibition of the ERK-dependent metalloprotease ADAM17, which normally inhibits c-MET signaling by promoting shedding of its endogenous antagonist, soluble "decoy" MET. Most importantly, pharmacological blockade of this resistance pathway with either c-MET or JAK1/2 inhibitors synergistically increased MEK-inhibitor-induced apoptosis and growth inhibition in vitro and in vivo in KRASMT models, providing clear rationales for the clinical assessment of these combinations in KRASMT CRC patients.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.