MilliporeSigma
  • Cannabinoid Type 2 Receptors Mediate a Cell Type-Specific Plasticity in the Hippocampus.
Cannabinoid Type 2 Receptors Mediate a Cell Type-Specific Plasticity in the Hippocampus.
Neuron (2016-05-03)
A Vanessa Stempel, Alexander Stumpf, Hai-Ying Zhang, Tuğba Özdoğan, Ulrike Pannasch, Anne-Kathrin Theis, David-Marian Otte, Alexandra Wojtalla, Ildikó Rácz, Alexey Ponomarenko, Zheng-Xiong Xi, Andreas Zimmer, Dietmar Schmitz
ABSTRACT
Endocannabinoids (eCBs) exert major control over neuronal activity by activating cannabinoid receptors (CBRs). The functionality of the eCB system is primarily ascribed to the well-documented retrograde activation of presynaptic CB1Rs. We find that action potential-driven eCB release leads to a long-lasting membrane potential hyperpolarization in hippocampal principal cells that is independent of CB1Rs. The hyperpolarization, which is specific to CA3 and CA2 pyramidal cells (PCs), depends on the activation of neuronal CB2Rs, as shown by a combined pharmacogenetic and immunohistochemical approach. Upon activation, they modulate the activity of the sodium-bicarbonate co-transporter, leading to a hyperpolarization of the neuron. CB2R activation occurred in a purely self-regulatory manner, robustly altered the input/output function of CA3 PCs, and modulated gamma oscillations in vivo. To conclude, we describe a cell type-specific plasticity mechanism in the hippocampus that provides evidence for the neuronal expression of CB2Rs and emphasizes their importance in basic neuronal transmission.
Sigma-Aldrich
Milli-Mark® Anti-NeuN-PE Antibody, clone A60, clone A60, Milli-Mark®, from mouse