MilliporeSigma
PGE(2) in the regulation of programmed erythrocyte death.
Cell death and differentiation (2005-03-05)
P A Lang, D S Kempe, S Myssina, V Tanneur, C Birka, S Laufer, F Lang, T Wieder, S M Huber
ABSTRACT
Hyperosmotic shock, energy depletion, or removal of extracellular Cl(-) activates Ca(2+)-permeable cation channels in erythrocyte membranes. Subsequent Ca(2+) entry induces erythrocyte shrinkage and exposure of phosphatidylserine (PS) at the erythrocyte surface. PS-exposing cells are engulfed by macrophages. The present study explored the signalling involved. Hyperosmotic shock and Cl(-) removal triggered the release of prostaglandin E(2) (PGE(2)). In whole-cell recording, activation of the cation channels by Cl(-) removal was abolished by the cyclooxygenase inhibitor diclophenac. In FACS analysis, phospholipase-A(2) inhibitors quinacrine and palmitoyltrifluoromethyl-ketone, and cyclooxygenase inhibitors acetylsalicylic acid and diclophenac, blunted the increase of PS exposure following Cl(-) removal. PGE(2) (but not thromboxane) induced cation channel activation, increase in cytosolic Ca(2+) concentration, cell shrinkage, PS exposure, calpain activation, and ankyrin-R degradation. The latter was attenuated by calpain inhibitors-I/II, while PGE(2)-induced PS exposure was not. In conclusion, hyperosmotic shock or Cl(-) removal stimulates erythrocyte PS exposure through PGE(2) formation and subsequent activation of Ca(2+)-permeable cation channels.
Sigma-Aldrich
Calpain Inhibitor II, powder
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
Anti-Ankyrin Antibody, clone Ank016, clone Ank016, Chemicon®, from mouse