MilliporeSigma
  • LncRNA BC088259 promotes Schwann cell migration through Vimentin following peripheral nerve injury.
LncRNA BC088259 promotes Schwann cell migration through Vimentin following peripheral nerve injury.
Glia (2019-11-14)
Chun Yao, Yanping Chen, Jing Wang, Tianmei Qian, Wei Feng, Yuanyuan Chen, Susu Mao, Bin Yu
ABSTRACT
Schwann cell, the major glial cell in the peripheral nervous system, plays an essential role in peripheral nerve regeneration. However, the regulation of Schwann cell behavior following nerve injury is insufficiently explored. According to the development of high-throughput techniques, long noncoding RNAs (lncRNAs) have been recognized. Accumulating evidence shows that lncRNAs take part in diverse biological processes and diseases. Here, by microarray analysis, we identified an upregulated lncRNA profile following sciatic nerve injury and focused on BC088259 for further investigation. Silencing or overexpression of BC088259 could affect Schwann cell migration. Mechanistically, BC088259 might exert this regulatory role by directly binding with Vimentin. Collectively, our study not only revealed a set of upregulated lncRNAs following nerve injury but also identified a new functional lncRNA, BC088259, which was important for Schwann cell migration, providing a therapeutic avenue toward peripheral nerve injury.
Sigma-Aldrich
MISSION® esiRNA, targeting human VIM