MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Live-cell imaging and ultrastructural analysis reveal remarkable features of cultured porcine gonocytes.
Live-cell imaging and ultrastructural analysis reveal remarkable features of cultured porcine gonocytes.
Cell and tissue research (2020-05-11)
Awang Hazmi Awang-Junaidi, Mohammad Amin Fayaz, Eiko Kawamura, LaRhonda Sobchishin, Daniel J MacPhee, Ali Honaramooz
ABSTRACT
Gonocytes in the neonatal testis have male germline stem cell potential. The objective of the present study was to examine the behavior and ultrastructure of gonocytes in culture. Neonatal porcine testis cells were cultured for 4 weeks and underwent live-cell imaging to explore real-time interactions among cultured cells. This included imaging every 1 h from day 0 to day 3, every 2 h from day 4 to day 7, and every 1 h for 24 h at days 14, 21, and 28. Samples also underwent scanning electron microscopy, transmission electron microscopy, morphometric evaluations, immunofluorescence, and RT-PCR. Live-cell imaging revealed an active amoeboid-like movement of gonocytes, assisted by the formation of extensive cytoplasmic projections, which, using scanning electron microscopy, were categorized into spike-like filopodia, leaf-like lamellipodia, membrane ruffles, and cytoplasmic blebs. In the first week of culture, gonocytes formed loose attachments on top of a somatic cell monolayer and, in week 2, formed grape-like clusters, which, over time, grew in cell number. Starting at week 3 of culture, some of the gonocyte clusters transformed into large multinucleated embryoid body-like colonies (EBLCs) that expressed both gonocyte- and pluripotent-specific markers. The number and diameter of individual gonocytes, the number and density of organelles within gonocytes, as well as the number and diameter of the EBLCs increased over time (P < 0.05). In conclusion, cultured porcine gonocytes displayed extensive migratory behavior facilitated by their various cytoplasmic projections, propagated, and transformed into EBLCs that increased in size and complexity over time.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%