MilliporeSigma
  • Early and natural embryonic death in Lagostomus maximus: Association with the uterine glands, vasculature, and musculature.
Early and natural embryonic death in Lagostomus maximus: Association with the uterine glands, vasculature, and musculature.
Journal of morphology (2020-05-08)
Francisco Acuña, Claudio G Barbeito, Enrique L Portiansky, Guadalupe Ranea, Fabian Nishida, María A Miglino, Mirta A Flamini
ABSTRACT
The uterus is an organ with great plasticity due to the morphological and physiological changes it experiences during the estrous cycle and pregnancy. In mammals, pregnancy requires diverse sex hormones, growth factors and cytokines, among others, for promoting uterine remodeling to favor implantation, placentation, and embryo/fetus survival and growth. The hystricognathi rodent Lagostomus maximus (plains viscacha) has a high rate of embryonic resorption. The cranial and middle implants are reabsorbed 25-35 days after intercourse while the caudal embryos continue with their development until two precocial offspring are born. So far, no uterine studies of non-pregnant L. maximus females were performed to determine the possible existence of variations in the organ that could be related to the differential survival of the implants. We used ultrasonography, as well as morphological, morphometric, histochemical, lectinhistochemical, and immunohistochemical methods to study differences in the uterine glands (area), vasculature (area), and musculature (thickness) along the uterine horns in non-pregnant females. Along the uterus, all these structures were in more advanced developmental condition in the caudal region as compared to more anterior positions. These regional variations could be decisive in explaining the reason why only caudal implantations come to term. In contrast, no differences in the in the luminal and glandular epithelial cells, nor in the degree of cell proliferation and apoptosis, and hormonal receptor staining were found. These parameters could be related to implantation along the uterine horns, but not to the differential survival of the implants.
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit