MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Phospholipase D activation is an early component of the salicylic acid signaling pathway in Arabidopsis cell suspensions.
Phospholipase D activation is an early component of the salicylic acid signaling pathway in Arabidopsis cell suspensions.
Plant physiology (2009-03-24)
Ondrej Krinke, Matyás Flemr, Chantal Vergnolle, Sylvie Collin, Jean-Pierre Renou, Ludivine Taconnat, Agnès Yu, Lenka Burketová, Olga Valentová, Alain Zachowski, Eric Ruelland
ABSTRACT
Salicylic acid (SA) plays a central role in defense against pathogen attack, as well as in germination, flowering, senescence, and the acquisition of thermotolerance. In this report we investigate the involvement of phospholipase D (PLD) in the SA signaling pathway. In presence of exogenous primary alcohols, the production of phosphatidic acid by PLD is diverted toward the formation of phosphatidylalcohols through a reaction called transphosphatidylation. By in vivo metabolic phospholipid labeling with (33)P(i), PLD activity was found to be induced 45 min after addition of SA. We show that incubation of Arabidopsis (Arabidopsis thaliana) cell suspensions with primary alcohols inhibited the induction of two SA-responsive genes, PATHOGENESIS-RELATED1 and WRKY38, in a dose-dependent manner. This inhibitory effect was more pronounced when the primary alcohols were more hydrophobic. Secondary or tertiary alcohols had no inhibitory effect. These results provide compelling arguments for PLD activity being upstream of the induction of these genes by SA. A subsequent study of n-butanol effects on the SA-responsive transcriptome identified 1,327 genes differentially expressed upon SA treatment. Strikingly, the SA response of 380 of these genes was inhibited by n-butanol but not by tert-butanol. A detailed analysis of the regulation of these genes showed that PLD could act both positively and negatively, either on gene induction or gene repression. The overlap with the previously described phosphatidylinositol-4-kinase pathway is discussed.
Sigma-Aldrich
DNase I, Amplification Grade
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
TRI Reagent®
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.