MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Negative regulation of interferon-induced transmembrane protein 3 by SET7-mediated lysine monomethylation.
Negative regulation of interferon-induced transmembrane protein 3 by SET7-mediated lysine monomethylation.
The Journal of biological chemistry (2013-10-17)
Zhao Shan, Qinglin Han, Jia Nie, Xuezhi Cao, Zuojia Chen, Shuying Yin, Yayi Gao, Fang Lin, Xiaohui Zhou, Ke Xu, Huimin Fan, Zhikang Qian, Bing Sun, Jin Zhong, Bin Li, Andy Tsun
ABSTRACT
Although lysine methylation is classically known to regulate histone function, its role in modulating antiviral restriction factor activity remains uncharacterized. Interferon-induced transmembrane protein 3 (IFITM3) was found monomethylated on its lysine 88 residue (IFITM3-K88me1) to reduce its antiviral activity, mediated by the lysine methyltransferase SET7. Vesicular stomatitis virus and influenza A virus infection increased IFITM3-K88me1 levels by promoting the interaction between IFITM3 and SET7, suggesting that this pathway could be hijacked to support infection; conversely, IFN-α reduced IFITM3-K88me1 levels. These findings may have important implications in the design of therapeutics targeting protein methylation against infectious diseases.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Rabbit IgG−Agarose, saline suspension