MilliporeSigma
  • Defactinib attenuates osteoarthritis by inhibiting positive feedback loop between H-type vessels and MSCs in subchondral bone.
Defactinib attenuates osteoarthritis by inhibiting positive feedback loop between H-type vessels and MSCs in subchondral bone.
Journal of orthopaedic translation (2020-06-11)
Yanjun Hu, Hangtian Wu, Ting Xu, Yutian Wang, Hanjun Qin, Zilong Yao, Peisheng Chen, Yongheng Xie, Zhiguo Ji, Kaifan Yang, Yu Chai, Xianrong Zhang, Bin Yu, Zhuang Cui
ABSTRACT
Abnormal bone formation in subchondral bone resulting from uncoupled bone remodeling is considered a central feature in osteoarthritis (OA) pathogenesis. H-type vessels can couple angiogenesis and osteogenesis. We previously revealed that elevated H-type vessels in subchondral bone were correlated with OA and focal adhesion kinase (FAK) in MSCs is critical for H-type vessel formation in osteoporosis. The aim of this study was to explore the correlation between H-type vessels and MSCs in OA pathogenesis through regulation of H-type vessel formation using defactinib (an FAK inhibitor). In vivo: 3-month-old male C57BL/6J (WT) mice were randomly divided into three groups: sham controls, vehicle-treated ACLT mice, and defactinib-treated ACLT mice (25 mg/kg, intraperitoneally weekly). In vitro: we explored the role of conditioned medium (CM) of MSCs from subchondral bone of different groups on the angiogenesis of endothelial cells (ECs). Flow cytometry, Western blotting, ELISA, real time (RT)-PCR, immunostaining, CT-based microangiography, and bone micro-CT (μCT) were used to detect changes in relative cells and tissues. This study demonstrated that inhibition of H-type vessels with defactinib alleviated OA by inhibiting H-type vessel-linked MSCs in subchondral bone. During OA pathogenesis, H-type vessels and MSCs formed a positive feedback loop contributing to abnormal bone formation in subchondral bone. Elevated H-type vessels provided indispensable MSCs for abnormal bone formation in subchondral bone. Flow cytometry and immunostaining results confirmed that the amount of MSCs in subchondral bone was obviously higher in vehicle-treated ACLT mice than that in sham controls and defactinib-treated ACLT mice. In vitro, p-FAK in MSCs from subchondral bone of vehicle-treated ALCT mice increased significantly relative to other groups. Further, the CM from MSCs of vehicle-treated ACLT mice enhanced angiogenesis of ECs through FAK-Grb2-MAPK-linked VEGF expression. Our results demonstrate that defactinib inhibits OA by suppressing the positive feedback loop between H-type vessels and MSCs in subchondral bone. Our results provide a mechanistic rationale for the use of defactinib as an effective candidate for OA treatment.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.