MilliporeSigma
  • Depression-like behaviors induced by defective PTPRT activity through dysregulated synaptic functions and neurogenesis.
Depression-like behaviors induced by defective PTPRT activity through dysregulated synaptic functions and neurogenesis.
Journal of cell science (2020-09-18)
So-Hee Lim, Sangyep Shin, Myoung-Hwan Kim, Eung Chang Kim, Da Yong Lee, Jeonghee Moon, Hye-Yeon Park, Young-Kyoung Ryu, Young-Mi Kang, Yu Jeong Kang, Tae Hwan Kim, Na-Yoon Lee, Nam-Soon Kim, Dae-Yeul Yu, Insop Shim, Yoichi Gondo, Masanobu Satake, Eunhee Kim, Kyoung-Shim Kim, Sun Seek Min, Jae-Ran Lee
ABSTRACT
PTPRT has been known to regulate synaptic formation and dendritic arborization of hippocampal neurons. PTPRT-/- null and PTPRT-D401A mutant mice displayed enhanced depression-like behaviors compared with wild-type mice. Transient knockdown of PTPRT in the dentate gyrus enhanced the depression-like behaviors of wild-type mice, whereas rescued expression of PTPRT ameliorated the behaviors of PTPRT-null mice. Chronic stress exposure reduced expression of PTPRT in the hippocampus of mice. In PTPRT-deficient mice the expression of GluR2 (also known as GRIA2) was attenuated as a consequence of dysregulated tyrosine phosphorylation, and the long-term potentiation at perforant-dentate gyrus synapses was augmented. The inhibitory synaptic transmission of the dentate gyrus and hippocampal GABA concentration were reduced in PTPRT-deficient mice. In addition, the hippocampal expression of GABA transporter GAT3 (also known as SLC6A11) was decreased, and its tyrosine phosphorylation was increased in PTPRT-deficient mice. PTPRT-deficient mice displayed reduced numbers and neurite length of newborn granule cells in the dentate gyrus and had attenuated neurogenic ability of embryonic hippocampal neural stem cells. In conclusion, our findings show that the physiological roles of PTPRT in hippocampal neurogenesis, as well as synaptic functions, are involved in the pathogenesis of depressive disorder.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid
Sigma-Aldrich
Anti-Parvalbumin Antibody, ascites fluid, clone PARV-19, Chemicon®