MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Subtle side-chain modifications of the hop phytoestrogen 8-prenylnaringenin result in distinct agonist/antagonist activity profiles for estrogen receptors alpha and beta.
Subtle side-chain modifications of the hop phytoestrogen 8-prenylnaringenin result in distinct agonist/antagonist activity profiles for estrogen receptors alpha and beta.
Journal of medicinal chemistry (2006-12-08)
Frederik Roelens, Nina Heldring, Willem Dhooge, Martin Bengtsson, Frank Comhaire, Jan-Ake Gustafsson, Eckardt Treuter, Denis De Keukeleire
ABSTRACT
In search of therapeutic agents for estrogen-related pathologies, phytoestrogens are being extensively explored. In contrast to naringenin, 8-prenylnaringenin is a potent hop-derived estrogenic compound, highlighting the importance of the prenyl group for hormonal activity. We investigated the effects of substituting the prenyl group at C(8) with alkyl chains of varying lengths and branching patterns on estrogen receptor (ER) subtype ERalpha- and ERbeta-binding affinities and transcriptional activities. In addition, features of the ligand-induced receptor conformations were explored using a set of specific ER-binding peptides. The new 8-alkylnaringenins were found to span an activity spectrum ranging from full agonism to partial agonism to antagonism. Most strikingly, 8-(2,2-dimethylpropyl)naringenin exhibited full agonist character on ERalpha, but pronounced antagonist character on ERbeta. Knowledge on how ER-subtype-selective activities can be designed provides valuable information for future drug or tool compound discovery.
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm, contains 2 nuts, 2 ferrules, 5cm tubing, 1 guard cartridge and 1 guard holder