MilliporeSigma
  • A small sustained increase in NOD1 abundance promotes ligand-independent inflammatory and oncogene transcriptional responses.
A small sustained increase in NOD1 abundance promotes ligand-independent inflammatory and oncogene transcriptional responses.
Science signaling (2020-12-10)
Leah M Rommereim, Ajay Suresh Akhade, Bhaskar Dutta, Carolyn Hutcheon, Nicolas W Lounsbury, Clifford C Rostomily, Ram Savan, Iain D C Fraser, Ronald N Germain, Naeha Subramanian, Leah M Rommereim, Ajay Suresh Akhade, Bhaskar Dutta, Carolyn Hutcheon, Nicolas W Lounsbury, Clifford C Rostomily, Ram Savan, Iain D C Fraser, Ronald N Germain, Naeha Subramanian
ABSTRACT
Small, genetically determined differences in transcription [expression quantitative trait loci (eQTLs)] are implicated in complex diseases through unknown molecular mechanisms. Here, we showed that a small, persistent increase in the abundance of the innate pathogen sensor NOD1 precipitated large changes in the transcriptional state of monocytes. A ~1.2- to 1.3-fold increase in NOD1 protein abundance resulting from loss of regulation by the microRNA cluster miR-15b/16 lowered the threshold for ligand-induced activation of the transcription factor NF-κB and the MAPK p38. An additional sustained increase in NOD1 abundance to 1.5-fold over basal amounts bypassed this low ligand concentration requirement, resulting in robust ligand-independent induction of proinflammatory genes and oncogenes. These findings reveal that tight regulation of NOD1 abundance prevents this sensor from exceeding a physiological switching checkpoint that promotes persistent inflammation and oncogene expression. Furthermore, our data provide insight into how a quantitatively small change in protein abundance can produce marked changes in cell state that can serve as the initiator of disease.
Sigma-Aldrich
Anti-ALX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution