MilliporeSigma
  • Synthesis and SAR of [1,2,4]triazolo[1,5-a]pyrimidines, a class of anticancer agents with a unique mechanism of tubulin inhibition.
Synthesis and SAR of [1,2,4]triazolo[1,5-a]pyrimidines, a class of anticancer agents with a unique mechanism of tubulin inhibition.
Journal of medicinal chemistry (2007-01-19)
Nan Zhang, Semiramis Ayral-Kaloustian, Thai Nguyen, Jay Afragola, Richard Hernandez, Judy Lucas, James Gibbons, Carl Beyer
ABSTRACT
The synthesis and SAR of a series of triazolopyrimidines as anticancer agents are described. Treatment of 5-chloro-6-(trifluorophenyl)-N-fluoroalkyl [1,2,4]triazolo[1,5-a]pyrimidin-7-amine with an alcohol, a thiol, or an alkylamine provided the corresponding final compounds. A clear SAR requirement has been established for optimal activity. A (1S)-2,2,2-trifluoro-1-methylethylamino group or an achiral 2,2,2-trifluoroethylamino group is required at the 5-position to achieve high potency. On the phenyl ring, both fluoro atoms, at the positions ortho to the triazolopyrimidine core, are needed for optimal activity. At the position para to the triazolopyrimidine core, on the phenyl ring, the best activity is achieved with an oxygen linkage followed by a three-methylene unit, and an alkylamino or a hydroxy group. The mechanism of action for this series of triazolopyrimidines was shown to be unique in that they promoted tubulin polymerization in vitro, but did not bind competitively with paclitaxel.1 Instead, they inhibit the binding of vincas to tubulin. Selected compounds were studied further, and it was shown that these compounds were able to overcome resistance attributed to several multidrug resistance transporter proteins. Lead compounds were shown to inhibit tumor growth in several nude mouse xenograft models, with high potency and efficacy, when dosed either orally or intravenously.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)