MilliporeSigma
  • Processed Eggshell Membrane Powder Is a Promising Biomaterial for Use in Tissue Engineering.
Processed Eggshell Membrane Powder Is a Promising Biomaterial for Use in Tissue Engineering.
International journal of molecular sciences (2020-11-05)
Sissel B Rønning, Ragnhild S Berg, Vibeke Høst, Eva Veiseth-Kent, Christian R Wilhelmsen, Eirik Haugen, Henri-Pierre Suso, Paul Barham, Ralf Schmidt, Mona E Pedersen
ABSTRACT
The purpose of this study was to investigate the tissue regenerating and biomechanical properties of processed eggshell membrane powder (PEP) for use in 3D-scaffolds. PEP is a low-cost, natural biomaterial with beneficial bioactive properties. Most importantly, this material is available as a by-product of the chicken egg processing (breaking) industry on a large scale, and it could have potential as a low-cost ingredient for therapeutic scaffolds. Scaffolds consisting of collagen alone and collagen combined with PEP were produced and analyzed for their mechanical properties and the growth of primary fibroblasts and skeletal muscle cells. Mechanical testing revealed that a PEP/collagen-based scaffold increased the mechanical hardness of the scaffold compared with a pure collagen scaffold. Scanning electron microscopy (SEM) demonstrated an interconnected porous structure for both scaffolds, and that the PEP was evenly distributed in dense clusters within the scaffold. Fibroblast and skeletal muscle cells attached, were viable and able to proliferate for 1 and 2 weeks in both scaffolds. The cell types retained their phenotypic properties expressing phenotype markers of fibroblasts (TE7, alpha-smooth muscle actin) and skeletal muscle (CD56) visualized by immunostaining. mRNA expression of the skeletal muscle markers myoD, myogenin, and fibroblasts marker (SMA) together with extracellular matrix components supported viable phenotypes and matrix-producing cells in both types of scaffolds. In conclusion, PEP is a promising low-cost, natural biomaterial for use in combination with collagen as a scaffold for 3D-tissue engineering to improve the mechanical properties and promote cellular adhesion and growth of regenerating cells.
Sigma-Aldrich
Anti-Fibroblasts Antibody, clone TE-7, clone TE-7, Chemicon®, from mouse