MilliporeSigma
  • Heterostructured CoOx-TiO2 Mesoporous/Photonic Crystal Bilayer Films for Enhanced Visible-Light Harvesting and Photocatalysis.
Heterostructured CoOx-TiO2 Mesoporous/Photonic Crystal Bilayer Films for Enhanced Visible-Light Harvesting and Photocatalysis.
Materials (Basel, Switzerland) (2020-10-01)
Stelios Loukopoulos, Alexia Toumazatou, Elias Sakellis, Evangelia Xenogiannopoulou, Nikos Boukos, Athanasios Dimoulas, Vlassis Likodimos
ABSTRACT
Heterostructured bilayer films, consisting of co-assembled TiO2 photonic crystals as the bottom layer and a highly performing mesoporous P25 titania as the top layer decorated with CoOx nanoclusters, are demonstrated as highly efficient visible-light photocatalysts. Broadband visible-light activation of the bilayer films was implemented by the surface modification of both titania layers with nanoscale clusters of Co oxides relying on the chemisorption of Co acetylacetonate complexes on TiO2, followed by post-calcination. Tuning the slow photon regions of the inverse opal supporting layer to the visible-light absorption of surface CoOx oxides resulted in significant amplification of salicylic-acid photodegradation under visible and ultraviolet (UV)-visible light (Vis), outperforming benchmark P25 films of higher titania loading. This enhancement was related to the spatially separated contributions of slow photon propagation in the inverse opal support layer assisted by Bragg reflection toward the CoOx-modified mesoporous P25 top layer. This effect indicates that photonic crystals may be highly effective as both photocatalytically active and backscattering layers in multilayer photocatalytic films.
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
α-Sexithiophene
Sigma-Aldrich
Titanium(IV) bis(ammonium lactato)dihydroxide solution, 50 wt. % in H2O
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)