MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Glucocorticoid receptor overexpression exerts an antisurvival effect on human small cell lung cancer cells.
Glucocorticoid receptor overexpression exerts an antisurvival effect on human small cell lung cancer cells.
Oncogene (2007-05-15)
P Sommer, P Le Rouzic, H Gillingham, A Berry, M Kayahara, T Huynh, A White, D W Ray
ABSTRACT
Small cell lung cancer (SCLC) is an aggressive tumour with an abysmal prognosis. These cancers are characteristically resistant to glucocorticoid (Gc) action, owing to impaired expression of the glucocorticoid receptor (GR). We identified reduced GR expression in human SCLC cell lines, compared to a non-SCLC cell line. The SCLC cells also showed no Gc inhibition of proliferation, in contrast to non-SCLC cells. Retroviral overexpression of GR resulted in significantly increased cell death, which was partially blocked by the GR antagonist, RU486. Indeed, in cells sorted for GR expression, there was rapid, near complete loss of live cells by 72 h, in contrast to control cells that proliferated as expected. Flow cytometry using Annexin V revealed that cell death was by apoptosis. In addition, confocal analysis of fixed cells showed that cells overexpressing GR displayed a significant increase in fragmenting apoptotic nuclei. Microarray studies showed that transgenic GR expression upregulated the proapoptotic genes, BAD and BAX. We have, therefore, identified a profound apoptotic effect of GR in SCLC cells, which may explain the low levels of endogenous GR in SCLC cells. Understanding how GR overexpression leads to apoptotic cell death in SCLC cells may uncover new therapeutic strategies.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid