MilliporeSigma
  • A novel cell adhesion inhibitor, K-7174, reduces the endothelial VCAM-1 induction by inflammatory cytokines, acting through the regulation of GATA.
A novel cell adhesion inhibitor, K-7174, reduces the endothelial VCAM-1 induction by inflammatory cytokines, acting through the regulation of GATA.
Biochemical and biophysical research communications (2000-06-02)
M Umetani, H Nakao, T Doi, A Iwasaki, M Ohtaka, T Nagoya, C Mataki, T Hamakubo, T Kodama
ABSTRACT
A novel inhibitor for the adhesion of monocytes to cytokine-stimulated endothelial cells, K-7174, was selected by an assay system using the cultured human monocytic cells and human endothelial cells. K-7174 inhibited the expression of vascular cell adhesion molecule-1 (VCAM-1) induced by either tumor necrosis factor alpha or interleukin-1beta, without affecting the induction of intercellular adhesion molecule-1 or E-selectin. K-7174 had no effect on the stability of VCAM-1 mRNA. Electrophoretic mobility shift assay revealed that its inhibitory effect on VCAM-1 induction was mediated by an effect on the binding to the GATA motifs in the VCAM-1 gene promoter region. K-7174 did not influence the binding to any of the following binding motifs: octamer binding protein, AP-1, SP-1, ets, NFkappaB, or interferon regulatory factor. These results suggest that the regulation of GATA binding may become a new target for anti-inflammatory drug development, acting through a mechanism independent from NFkappaB activity.
Sigma-Aldrich
K-7174 dihydrochloride, ≥98% (HPLC)