MilliporeSigma
  • PD-1+CD8+ T cells are clonally expanding effectors in human chronic inflammation.
PD-1+CD8+ T cells are clonally expanding effectors in human chronic inflammation.
The Journal of clinical investigation (2018-09-11)
Alessandra Petrelli, Gerdien Mijnheer, David P Hoytema van Konijnenburg, Maria M van der Wal, Barbara Giovannone, Enric Mocholi, Nadia Vazirpanah, Jasper C Broen, Dirkjan Hijnen, Bas Oldenburg, Paul J Coffer, Sebastian J Vastert, Berent J Prakken, Eric Spierings, Aridaman Pandit, Michal Mokry, Femke van Wijk
ABSTRACT
Chronic inflammatory diseases are characterized by recurrent inflammatory attacks in the tissues mediated by autoreactive T cells. Identity and functional programming of CD8+ T cells at the target site of inflammation still remain elusive. One key question is whether, in these antigen-rich environments, chronic stimulation leads to CD8+ T cell exhaustion comparable to what is observed in infectious disease contexts. In the synovial fluid (SF) of juvenile idiopathic arthritis (JIA) patients, a model of chronic inflammation, an overrepresentation of PD-1+CD8+ T cells was found. Gene expression profiling, gene set enrichment analysis, functional studies, and extracellular flux analysis identified PD-1+CD8+ T cells as metabolically active effectors, with no sign of exhaustion. Furthermore, PD-1+CD8+ T cells were enriched for a tissue-resident memory (Trm) cell transcriptional profile and demonstrated increased clonal expansion compared with the PD-1- counterpart, suggesting antigen-driven expansion of locally adapted cells. Interestingly, this subset was also found increased in target tissues in other human chronic inflammatory diseases. These data indicate that local chronic inflammation drives the induction and expansion of CD8+ T cells endowed with potential detrimental properties. Together, these findings lay the basis for investigation of PD-1-expressing CD8+ T cell targeting strategies in human chronic inflammatory diseases.
Sigma-Aldrich
AEC Staining Kit, liquid