MilliporeSigma
  • Novel 2,4,5-trisubstituted oxazole derivatives: synthesis and antiproliferative activity.
Novel 2,4,5-trisubstituted oxazole derivatives: synthesis and antiproliferative activity.
European journal of medicinal chemistry (2009-05-09)
Xin-Hua Liu, Peng-Cheng Lv, Jia-Yu Xue, Bao-An Song, Hai-Liang Zhu
ABSTRACT
Microwave irradiation promotes the rapid O,N-acylation-cyclodehydration cascade reaction of oximes and acid chloride. Twenty novel 2,4,5-trisubstituted oxazole derivatives containing heterocycle moiety were synthesized and evaluated for their antiproliferative activity. The twenty compounds are all first reported and their structures were established by elemental analysis, (1)H NMR and (13)C NMR spectra. The bioassay tests showed that compounds 2-(2-(2-fluorophenyl)-4-(2,3,4-trimethoxyphenyl)oxazol-5-ylthio)benzo[d]thiazole (6af), 2-(2-(pyridin-3-yl)-4-(2,3,4-trimethoxyphenyl)oxazol-5-ylthio)pyrimidine (6bg) and 2-(2-(2-fluorophenyl)-4-(2,3,4-trimethoxyphenyl)oxazol-5-ylthio)-5-methyl-1,3,4-thiadiazole (6cf) displayed good antiproliferative activity in vitro, which were comparable to the positive control (5-fluorouracil).
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications