MilliporeSigma
  • Isoflavonoids from Erythrina poeppigiana: evaluation of their binding affinity for the estrogen receptor.
Isoflavonoids from Erythrina poeppigiana: evaluation of their binding affinity for the estrogen receptor.
Journal of natural products (2009-08-27)
Sefirin Djiogue, Maria Halabalaki, Xanthippi Alexi, Dieudonne Njamen, Zacharias Tanee Fomum, Michael N Alexis, Alexios-Leandros Skaltsounis
ABSTRACT
Five new isoflavones, named 5,4'-dihydroxy-7-methoxy-3'-(3-methylbuten-2-yl)isoflavone (1), 5,2',4'-trihydroxy-7-methoxy-5'-(3-methylbuten-2-yl)isoflavone (2), 5,4'-dihydroxy-7-methoxy-3'-(3-methyl-2-hydroxybuten-3-yl)isoflavone (3), 3'-formyl-5,4'-dihydroxy-7-methoxyisoflavone (4), and 5-hydroxy-3''-hydroxy-2'',2''-dimethyldihydropyrano[5'',6'':3',4']isoflavone (5), as well as six known compounds, wighteone (6), 3'-isoprenylgenistein (7), isolupabigenin (8), alpinumisoflavone (9), erypoegin D (10), and crystacarpin (11), were isolated from Erythrina poeppigiana. The structures of the isolated compounds were elucidated on the basis of chemical and spectroscopic analysis. The affinity of these compounds for the estrogen receptors ERalpha and ERbeta was evaluated using a receptor binding assay. While isoprenyl and dimethylpyrano substituents in ring A reduced the affinity of binding to ERbeta ca. 100-fold compared to genistein, the isoprenyl substituent in ring B was better accommodated, allowing 7 to bind with ca. 10-fold lower affinity than genistein.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture