MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • From 6-aminoquinolone antibacterials to 6-amino-7-thiopyranopyridinylquinolone ethyl esters as inhibitors of Staphylococcus aureus multidrug efflux pumps.
From 6-aminoquinolone antibacterials to 6-amino-7-thiopyranopyridinylquinolone ethyl esters as inhibitors of Staphylococcus aureus multidrug efflux pumps.
Journal of medicinal chemistry (2010-05-08)
Marco Pieroni, Mirjana Dimovska, Jean Pierre Brincat, Stefano Sabatini, Emanuele Carosati, Serena Massari, Glenn W Kaatz, Arnaldo Fravolini
ABSTRACT
The thiopyranopyridine moiety was synthesized as a new heterocyclic base to be inserted at the C-7 position of selected quinolone nuclei followed by a determination of antibacterial activity against strains of Staphylococcus aureus. Selected thiopyranopyridinylquinolones showed significant antimicrobial activity, including strains having mutations in gyrA and grlA as well as other strains overexpressing the NorA multidrug (MDR) efflux pump. Most derivatives did not appear to be NorA substrates. The effect of the thiopyranopyridinyl substituent on making these quinolones poor substrates for NorA was investigated further. Several quinolone ester intermediates, devoid of any intrinsic antibacterial activity, were tested for their abilities to inhibit the activities of NorA (MFS family) and MepA (MATE family) S. aureus MDR efflux pumps. Selected quinolone esters were capable of inhibiting both MDR pumps more efficiently than the reference compound reserpine. Moreover, they also were able to restore, and even enhance, the activity of ciprofloxacin toward some genetically modified resistant S. aureus strains.
Sigma-Aldrich
6-Aminoquinoline, 98%
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)