MilliporeSigma
  • Synthesis, chiral resolution, and determination of novel furan lignan derivatives with potent anti-tumor activity.
Synthesis, chiral resolution, and determination of novel furan lignan derivatives with potent anti-tumor activity.
Bioorganic & medicinal chemistry letters (2010-02-23)
Hai Ling Sun, Tian Tian Wang, Zhi Liang Lv, Ji Lu Feng, Dong Ping Geng, Yong Mei Li, Ke Li
ABSTRACT
A kind of racemic furan lignans were synthesized via a novel route, and two optical isomers were obtained through a selective hydrolization. The absolute configurations were determined by circular dichroism spectroscopy after separated through a preparative column. The different activities of isomers and the racemates were evaluated on QGY-7701 and HeLa cell lines, and compound 2c showed the best activity on QGY-7701 and HeLa cell lines with IC(50) 12 microM and 13 microM, respectively.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder