MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Rapid bioprinting of conjunctival stem cell micro-constructs for subconjunctival ocular injection.
Rapid bioprinting of conjunctival stem cell micro-constructs for subconjunctival ocular injection.
Biomaterials (2020-11-01)
Zheng Zhong, Xiaoqian Deng, Pengrui Wang, Claire Yu, Wisarut Kiratitanaporn, Xiaokang Wu, Jacob Schimelman, Min Tang, Alis Balayan, Emmie Yao, Jing Tian, Luwen Chen, Kang Zhang, Shaochen Chen
ABSTRACT
Ocular surface diseases including conjunctival disorders are multifactorial progressive conditions that can severely affect vision and quality of life. In recent years, stem cell therapies based on conjunctival stem cells (CjSCs) have become a potential solution for treating ocular surface diseases. However, neither an efficient culture of CjSCs nor the development of a minimally invasive ocular surface CjSC transplantation therapy has been reported. Here, we developed a robust in vitro expansion method for primary rabbit-derived CjSCs and applied digital light processing (DLP)-based bioprinting to produce CjSC-loaded hydrogel micro-constructs for injectable delivery. Expansion medium containing small molecule cocktail generated fast dividing and highly homogenous CjSCs for more than 10 passages in feeder-free culture. Bioprinted hydrogel micro-constructs with tunable mechanical properties enabled the 3D culture of CjSCs while supporting viability, stem cell phenotype, and differentiation potency into conjunctival goblet cells. These hydrogel micro-constructs were well-suited for scalable dynamic suspension culture of CjSCs and were successfully delivered to the bulbar conjunctival epithelium via minimally invasive subconjunctival injection. This work integrates novel cell culture strategies with bioprinting to develop a clinically relevant injectable-delivery approach for CjSCs towards the stem cell therapies for the treatment of ocular surface diseases.
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%
Sigma-Aldrich
DMH1, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl phenylphosphonite, 97%
Sigma-Aldrich
Triton X-100, laboratory grade