MilliporeSigma
  • Key aroma compounds of Chinese dry-cured Spanish mackerel (Scomberomorus niphonius) and their potential metabolic mechanisms.
Key aroma compounds of Chinese dry-cured Spanish mackerel (Scomberomorus niphonius) and their potential metabolic mechanisms.
Food chemistry (2020-10-25)
Siliang Wu, Juan Yang, Hao Dong, Qiaoyu Liu, Xiangluan Li, Xiaofang Zeng, Weidong Bai
ABSTRACT
The key aroma compounds of six commercially available dry-cured Spanish mackerel (Scomberomorus niphonius, DCSM) were identified using electronic nose (E-nose), gas chromatography-olfactometry (GC-O), and two-dimensional gas chromatography-time-of-flight mass spectrometry (GC × GC-TOFMS). A total of 38-55 aroma compounds were identified, and 21-26 of them, which presented high flavor dilution factors based on aroma extract dilution analysis, were quantified. Lastly, 9-14 key aroma compounds with high odor-active value, including 3-methyl-1-butanal, octanal, 1-octen-3-ol, nonanal, cis-4-decenal, ethyl caproate, (E)-2-octenal, (Z)-2-nonenal decanal, 3-methyl-1-butanol, 1-heptanol, 3-octanone, 2-octanol, and 6-methyl-5-hepten-2-one, were identified as the key aroma contributors in DCSM. Results also indicated that a longer dry-curing time would promote the generation of aroma compounds. The metabolism analysis implied that the auto-oxidation/oxidation of unsaturated fatty acids, such as oleic and linoleic acid, and the enzymatic degradation of l-leucine might be potential metabolic mechanisms.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Octanal, 99%
Sigma-Aldrich
Ethyl hexanoate, ≥99%
Sigma-Aldrich
cis-4-Decenal, ≥93%, stabilized, FG
Sigma-Aldrich
Decanal, natural, ≥97%, FG