MilliporeSigma
  • Cyclophilin D counterbalances mitochondrial calcium uniporter-mediated brain mitochondrial calcium uptake.
Cyclophilin D counterbalances mitochondrial calcium uniporter-mediated brain mitochondrial calcium uptake.
Biochemical and biophysical research communications (2020-07-25)
Bei Zhang, Kun Jia, Jing Tian, Heng Du
ABSTRACT
Mitochondria play an essential role in maintaining intraneuronal calcium homeostasis. Mitochondrial calcium uniporter (MCU) is a determined major brain mitochondrial calcium entry pathway. Activated MCU-mediated mitochondrial calcium overloading has been linked with brain mitochondrial pathology in disease conditions. Cyclophilin D (CypD)-mediated mitochondrial permeability transition (mPT) favors mitochondrial calcium efflux. The physiological function of CypD-mediated mPT has received increasing recognition. However, the regulatory role of CypD-mediated mPT in brain mitochondrial calcium dynamics in response to mitochondrial calcium accumulation via MCU has not been comprehensively studied. Here, by adopting purified brain mitochondria, we have determined an effect of CypD and CypD-mediated mPT against mitochondrial calcium overloading. In addition, blockade of CypD pharmaceutically or genetically blunts brain mitochondrial MCU's sensitivity to its inhibitor. Therefore, our findings suggest that CypD-mediated mPT is a mitochondrial compensatory response to MCU-mediated excess mitochondrial calcium accumulation. Moreover, CypD may potentially modulate MCU function in calcium-stressed mitochondria.
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
Ru360, Ru360, is a cell-permeable oxygen-bridged dinuclear ruthenium amine complex. Binds to mitochondria with high affinity (Kd = 340 pM) and blocks Ca2+ uptake into mitochondria in vitro (IC50 = 184 pM).