MilliporeSigma
  • Antiosteoclastic bone resorption activity of osteoprotegerin via enhanced AKT/mTOR/ULK1-mediated autophagic pathway.
Antiosteoclastic bone resorption activity of osteoprotegerin via enhanced AKT/mTOR/ULK1-mediated autophagic pathway.
Journal of cellular physiology (2019-09-20)
Hongyan Zhao, Ziqiang Sun, Yonggang Ma, Ruilong Song, Yan Yuan, Jianchun Bian, Jianhong Gu, Zongping Liu
ABSTRACT
Autophagy plays a critical role in the maintenance of bone homeostasis. Osteoprotegerin (OPG) is an inhibitor of osteoclast-mediated bone resorption. However, whether autophagy is involved in the antiosteoclastogenic effects of OPG remains unclear. The present study aimed to investigate the potential mechanism of autophagy during OPG-induced bone resorption via inhibition of osteoclasts differentiated from bone marrow-derived macrophages in BALB/c mice. The results showed that after treatment with receptor activator of nuclear factor-κΒ ligand and macrophage colony-stimulating factor for 3 days, TRAP+ osteoclasts formed, representing the resting state of autophagy. These osteoclasts were treated with OPG and underwent autophagy, as demonstrated by LC3-II accumulation, acidic vesicular organelle formation, and the presence of autophagosomes. The levels of autophagy-related proteins, LC3-II increased and P62 decreased at 3 hr in OPG-treated osteoclasts. The viability, differentiation, and bone resorption activity of osteoclasts declined after OPG treatment. Treatment with OPG and chloroquine, an autophagy inhibitor, attenuated OPG-induced inhibition of osteoclastic bone resorption, whereas rapamycin (RAP), an autophagy inducer, enhanced OPG-induced inhibition of differentiation, survival, and bone resorption activity of osteoclasts. Furthermore, OPG reduced the amount of phosphorylated(p) protein kinase B (AKT) and pmTOR and increased the level of pULK, in a dose-dependant manner. LY294002, a phosphatidylinositol-4,5-bisphosphate 3-kinase (PI3K)/AKT pathway inhibitor, attenuated the decline in pAKT, but enhanced the decline in pmTOR and the increase in pULK1 following OPG treatment. RAP enhanced the OPG-induced increase in pULK1. The PI3K inhibitor 3-methyladenine partly blocked OPG-induced autophagy. Thus, the results revealed that OPG inhibits osteoclast bone resorption by inducing autophagy via the AKT/mTOR/ULK1 signaling pathway.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor