MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of lactic fermentation on soy protein digestive pattern assessed by an in vitro dynamic gastrointestinal digestion model and the influence on human faecal microbiota.
Effect of lactic fermentation on soy protein digestive pattern assessed by an in vitro dynamic gastrointestinal digestion model and the influence on human faecal microbiota.
Journal of the science of food and agriculture (2020-07-31)
Ziyu Zhang, Yumeng Fu, Qi Liu, Benjamin K Simpson, Elham Azarpazhooh, Mingsheng Dong, Xin Rui
ABSTRACT
The aim of this study was to investigate the effect of lactic fermentation on soy protein gastrointestinal digestive pattern and the influence of protein digesta on human faecal microbiota. Soymilk and soy yogurt were prepared in this study and a novel in vitro dynamic gastrointestinal model was employed to simulate gastric and duodenum digestions. Particle size, sodium dodecyl sulphate polyacrylamide gel electrophoresis (SDS-PAGE), and peptide content were monitored at the end of duodenum tract. Ingestion of soy yogurt allowed a rapid drop in pH from 7.0 to 5.0 at simulated duodenal digestion (0-30 min), and resulted in a loss in soluble protein content compared to that of soymilk. The electrophoretic pattern between soymilk and soy yogurt exerted distinctive differences at early stages of duodenal digestion (0-60 min) and resulted in different peptide contents (180 min). Soy yogurt duodenal digesta collected at 180 min (D180), by co-fermentation with human intestinal flora distribution, allowed a higher population in Bifidobacterium spp., Lactobacillus/Enterococcus spp. and Streptococcus/Lactococcus spp., whereas soy yogurt D30 resulted in lower population in Clostridium and Escherichia coli compared to samples co-fermented with soymilk digesta. The results demonstrated lactic fermentation of soy protein modulated human intestinal microflora and might relate to the different protein digestive behaviours. © 2020 Society of Chemical Industry.
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Folin & Ciocalteu′s phenol reagent, suitable for determination of total protein by Lowry method, 1.9-2.1 N
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Mucin from porcine stomach, Type II
Millipore
Bile salts, suitable for microbiology